Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243802.1 | NP_001230731.1 | 4 - 5 | 351 | 86 | |
GenBank mRNA | AK345213.1 | - | 4 - 5 | 351 | 86 | |
AK346133.1 | - | 4 - 5 | 351 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |