Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243821.1 | NP_001230750.1 | 5 - 6 | 5322 | 102 | |
XM_005664006.3 | XP_005664063.1 | 5360 | 102 | |||
GenBank mRNA | CN155723.1 | - | 2 - 3 | 535 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |