Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243825.1 | NP_001230754.1 | 2 - 3 | 185 | 89 | |
GenBank mRNA | AK232666.1 | - | 2 - 3 | 177 | 89 | |
AK236961.1 | - | 2 - 3 | 185 | 89 | ||
AK343873.1 | - | 2 - 3 | 200 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |