Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC66A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243851.1 | NP_001230780.1 | 5 - 6 | 636 | 68 | |
GenBank mRNA | AK240018.1 | - | 5 - 6 | 636 | 68 | |
AK351824.1 | - | 2 - 3 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |