Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM32A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243887.1 | NP_001230816.1 | 2 - 3 | 243 | 97 | |
GenBank mRNA | AK239819.1 | - | 2 - 3 | 243 | 97 | |
AK395834.1 | - | 2 - 3 | 255 | 97 | ||
AK400888.1 | - | 2 - 3 | 243 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |