Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243896.1 | NP_001230825.1 | 2 - 3 | 233 | 77 | |
GenBank mRNA | AK234631.1 | - | 2 - 3 | 227 | 77 | |
AK238735.1 | - | 2 - 3 | 233 | 77 | ||
AK239831.1 | - | 2 - 3 | 215 | 77 | ||
AK347564.1 | - | 2 - 3 | 227 | 77 | ||
AK392606.1 | - | 2 - 3 | 224 | 77 | ||
AY609618.1 | - | 2 - 3 | 207 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |