Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243896.1 | NP_001230825.1 | 9 - 10 | 889 | 72 | |
GenBank mRNA | AK234631.1 | - | 9 - 10 | 883 | 72 | |
AK238735.1 | - | 9 - 10 | 889 | 72 | ||
AK347564.1 | - | 9 - 10 | 883 | 72 | ||
AK392606.1 | - | 9 - 10 | 72 | |||
AY609618.1 | - | 9 - 10 | 863 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |