Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243905.1 | NP_001230834.1 | 3 - 4 | 277 | 89 | |
GenBank mRNA | AK232815.1 | - | 3 - 4 | 277 | 89 | |
AY609754.1 | - | 3 - 4 | 276 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |