Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243920.1 | NP_001230849.1 | 5 - 6 | 560 | 94 | |
GenBank mRNA | AK231775.1 | - | 5 - 6 | 560 | 94 | |
AK391890.1 | - | 5 - 6 | 627 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |