Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243921.1 | NP_001230850.1 | 4 - 5 | 338 | 69 | |
XM_013981248.2 | XP_013836702.1 | 890 | 69 | |||
GenBank mRNA | AK237132.1 | - | 4 - 5 | 342 | 69 | |
AK240304.1 | - | 4 - 5 | 338 | 69 | ||
AK398479.1 | - | 4 - 5 | 715 | 69 | ||
AY609605.1 | - | 4 - 5 | 335 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |