Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243928.1 | NP_001230857.1 | 7 - 8 | 935 | 99 | |
XM_005656989.3 | XP_005657046.1 | 99 | ||||
GenBank mRNA | AK235509.1 | - | 7 - 8 | 99 | ||
AK236207.1 | - | 7 - 8 | 935 | 99 | ||
AK351448.1 | - | 5 - 6 | 99 | |||
AK390079.1 | - | 7 - 8 | 99 | |||
AK400049.1 | - | 7 - 8 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |