Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243934.1 | NP_001230863.1 | 2 - 3 | 253 | 60 | |
XM_021102014.1 | XP_020957673.1 | 1208 | 60 | |||
XM_021102015.1 | XP_020957674.1 | 1411 | 60 | |||
XM_021102017.1 | XP_020957676.1 | 1063 | 60 | |||
XM_021102018.1 | XP_020957677.1 | 1208 | 60 | |||
XM_021102019.1 | XP_020957678.1 | 1063 | 60 | |||
GenBank mRNA | AK349243.1 | - | 3 - 4 | 310 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |