Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SET | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244090.1 | NP_001231019.1 | 4 - 5 | 585 | 146 | |
XM_021071997.1 | XP_020927656.1 | 535 | 146 | |||
XM_021072036.1 | XP_020927695.1 | 533 | 146 | |||
XM_021072067.1 | XP_020927726.1 | 736 | 146 | |||
GenBank mRNA | AK347372.1 | - | 4 - 5 | 585 | 146 | |
AK351041.1 | - | 4 - 5 | 496 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |