Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244104.1 | NP_001231033.1 | 5 - 6 | 913 | 57 | |
XM_005661448.1 | XP_005661505.1 | 1248 | 57 | |||
XM_005661449.3 | XP_005661506.1 | 1088 | 57 | |||
GenBank mRNA | AK232358.1 | - | 4 - 5 | 705 | 57 | |
AK234563.1 | - | 5 - 6 | 912 | 57 | ||
AK235641.1 | - | 5 - 6 | 579 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |