Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244155.1 | NP_001231084.1 | 1 - 2 | 443 | 83 | |
XM_005666776.2 | XP_005666833.1 | 438 | 83 | |||
GenBank mRNA | AK236346.1 | - | 1 - 2 | 430 | 83 | |
AK395497.1 | - | 1 - 2 | 443 | 83 | ||
AY609583.1 | - | 1 - 2 | 242 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |