Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244176.1 | NP_001231105.1 | 4 - 5 | 533 | 71 | |
XM_021078526.1 | XP_020934185.1 | 561 | 71 | |||
XM_021078530.1 | XP_020934189.1 | 561 | 71 | |||
GenBank mRNA | AK231938.1 | - | 4 - 5 | 518 | 71 | |
AK234688.1 | - | 4 - 5 | 470 | 71 | ||
AK348653.1 | - | 4 - 5 | 472 | 71 | ||
AK398750.1 | - | 4 - 5 | 547 | 71 | ||
AY610152.1 | - | 4 - 5 | 533 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |