Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244188.1 | NP_001231117.1 | 2 - 3 | 150 | 71 | |
XM_021097994.1 | XP_020953653.1 | 397 | 71 | |||
XM_021097995.1 | XP_020953654.1 | 193 | 71 | |||
GenBank mRNA | AK231107.1 | - | 3 - 4 | 241 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |