Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244188.1 | NP_001231117.1 | 4 - 5 | 3606 | 73 | |
XM_005656279.3 | XP_005656336.1 | 5338 | 73 | |||
XM_021097992.1 | XP_020953651.1 | 4016 | 73 | |||
XM_021097993.1 | XP_020953652.1 | 4116 | 73 | |||
XM_021097994.1 | XP_020953653.1 | 3853 | 73 | |||
XM_021097995.1 | XP_020953654.1 | 3649 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |