Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244207.1 | NP_001231136.1 | 1 - 2 | 152 | 70 | |
XM_021085729.1 | XP_020941388.1 | 195 | 70 | |||
XM_021085730.1 | XP_020941389.1 | 195 | 70 | |||
GenBank mRNA | AK236724.1 | - | 1 - 2 | 152 | 70 | |
AK395930.1 | - | 1 - 2 | 195 | 70 | ||
AK398214.1 | - | 1 - 2 | 152 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |