Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244207.1 | NP_001231136.1 | 8 - 9 | 1097 | 75 | |
XM_005662769.3 | XP_005662826.1 | 1056 | 75 | |||
XM_013996366.2 | XP_013851820.1 | 1402 | 75 | |||
XM_021085729.1 | XP_020941388.1 | 972 | 75 | |||
XM_021085730.1 | XP_020941389.1 | 1007 | 75 | |||
XM_021085731.1 | XP_020941390.1 | 922 | 75 | |||
GenBank mRNA | AK236724.1 | - | 8 - 9 | 1097 | 75 | |
AK395930.1 | - | 8 - 9 | 1140 | 75 | ||
AK398214.1 | - | 8 - 9 | 1097 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |