Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BFAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244218.1 | NP_001231147.1 | 2 - 3 | 518 | 63 | |
XM_021085723.1 | XP_020941382.1 | 775 | 63 | |||
XM_021085724.1 | XP_020941383.1 | 904 | 63 | |||
XM_021085725.1 | XP_020941384.1 | 683 | 63 | |||
XM_021085726.1 | XP_020941385.1 | 769 | 63 | |||
XM_021085727.1 | XP_020941386.1 | 769 | 63 | |||
GenBank mRNA | AK239225.1 | - | 2 - 3 | 518 | 63 | |
AK352503.1 | - | 2 - 3 | 264 | 63 | ||
AK400735.1 | - | 3 - 4 | 608 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |