Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZXDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244222.1 | NP_001231151.1 | 4 - 5 | 1264 | 70 | |
XM_013996783.2 | XP_013852237.1 | 1325 | 70 | |||
GenBank mRNA | CN155436.1 | - | 3 - 4 | 363 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |