Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244232.1 | NP_001231161.1 | 1 - 2 | 72 | 85 | |
GenBank mRNA | BF190116.1 | - | 1 - 2 | 52 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |