Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244232.1 | NP_001231161.1 | 4 - 5 | 473 | 83 | |
GenBank mRNA | AK231186.1 | - | 3 - 4 | 686 | 83 | |
DY423750.1 | - | 2 - 3 | 252 | 83 | ||
EW092492.2 | - | 3 - 4 | 346 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |