Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244240.1 | NP_001231169.1 | 1 - 2 | 86 | 94 | |
GenBank mRNA | AK231622.1 | - | 1 - 2 | 66 | 94 | |
AK236902.1 | - | 1 - 2 | 69 | 94 | ||
AK237374.1 | - | 1 - 2 | 102 | 94 | ||
AK396003.1 | - | 1 - 2 | 69 | 94 | ||
FD635764.1 | - | 1 - 2 | 90 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |