Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244243.2 | NP_001231172.1 | 3 - 4 | 604 | 107 | |
XM_005669556.3 | XP_005669613.1 | 107 | ||||
XM_005669557.3 | XP_005669614.1 | 107 | ||||
XM_005669558.3 | XP_005669615.1 | 107 | ||||
XM_021068476.1 | XP_020924135.1 | 107 | ||||
XM_021068477.1 | XP_020924136.1 | 107 | ||||
XM_021068478.1 | XP_020924137.1 | 107 | ||||
XM_021068479.1 | XP_020924138.1 | 107 | ||||
XM_021068480.1 | XP_020924139.1 | 107 | ||||
XM_021068481.1 | XP_020924140.1 | 107 | ||||
XM_021068482.1 | XP_020924141.1 | 107 | ||||
XM_021068483.1 | XP_020924142.1 | 107 | ||||
XM_021068484.1 | XP_020924143.1 | 107 | ||||
XM_021068485.1 | XP_020924144.1 | 107 | ||||
XM_021068486.1 | XP_020924145.1 | 107 | ||||
XM_021068487.1 | XP_020924146.1 | 107 | ||||
GenBank mRNA | AK234347.1 | - | 3 - 4 | 605 | 107 | |
AK391715.1 | - | 3 - 4 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |