Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC33A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244245.1 | NP_001231174.1 | 3 - 4 | 1567 | 82 | |
XM_005669929.3 | XP_005669986.1 | 1599 | 82 | |||
XM_021068489.1 | XP_020924148.1 | 1649 | 82 | |||
GenBank mRNA | AK349530.1 | - | 3 - 4 | 1566 | 82 | |
AK400803.1 | - | 3 - 4 | 1567 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |