Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244248.1 | NP_001231177.1 | 2 - 3 | 744 | 84 | |
XM_021064040.1 | XP_020919699.1 | 881 | 84 | |||
GenBank mRNA | AK393378.1 | - | 3 - 4 | 821 | 84 | |
AY609415.1 | - | 3 - 4 | 794 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |