Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244270.1 | NP_001231199.1 | 2 - 3 | 479 | 81 | |
XM_005659660.1 | XP_005659717.1 | 492 | 81 | |||
XM_005659663.2 | XP_005659720.1 | 393 | 81 | |||
XM_005659664.1 | XP_005659721.1 | 492 | 81 | |||
XM_005659666.1 | XP_005659723.1 | 492 | 81 | |||
XM_013993066.1 | XP_013848520.1 | 492 | 81 | |||
XM_021074414.1 | XP_020930073.1 | 492 | 81 | |||
GenBank mRNA | HQ654520.1 | - | 2 - 3 | 479 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |