Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THUMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244282.1 | NP_001231211.1 | 1 - 2 | 340 | 67 | |
XM_005662087.3 | XP_005662144.1 | 674 | 67 | |||
XM_013995606.2 | XP_013851060.1 | 676 | 67 | |||
GenBank mRNA | AK235689.1 | - | 1 - 2 | 328 | 67 | |
AK395562.1 | - | 1 - 2 | 340 | 67 | ||
FS688700.1 | - | 1 - 2 | 340 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |