Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244308.1 | NP_001231237.1 | 1 - 2 | 148 | 88 | |
GenBank mRNA | AK235790.1 | - | 1 - 2 | 173 | 88 | |
AK237436.1 | - | 1 - 2 | 177 | 88 | ||
AK347757.1 | - | 1 - 2 | 148 | 88 | ||
FS692729.1 | - | 1 - 2 | 148 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |