Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244330.1 | NP_001231259.1 | 6 - 7 | 722 | 61 | |
XM_021066151.1 | XP_020921810.1 | 813 | 61 | |||
GenBank mRNA | AK234937.1 | - | 6 - 7 | 726 | 61 | |
AK236764.1 | - | 6 - 7 | 722 | 61 | ||
AK237732.1 | - | 4 - 5 | 654 | 61 | ||
AK345424.1 | - | 6 - 7 | 696 | 61 | ||
AK391275.1 | - | 6 - 7 | 724 | 61 | ||
AK396108.1 | - | 6 - 7 | 818 | 61 | ||
AK397757.1 | - | 7 - 8 | 798 | 61 | ||
KF501048.1 | - | 6 - 7 | 724 | 61 | ||
KU358546.1 | - | 5 - 6 | 670 | 61 |
Gene Symbol: LOC110255214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021067114.1 | XP_020922773.1 | 1054 | 61 | ||
XM_021067115.1 | XP_020922774.1 | 1054 | 61 | |||
XM_021067116.1 | XP_020922775.1 | 1203 | 61 | |||
XM_021067117.1 | XP_020922776.1 | 823 | 61 | |||
XM_021067118.1 | XP_020922777.1 | 1054 | 61 | |||
XM_021067120.1 | XP_020922779.1 | 823 | 61 | |||
GenBank mRNA | AK396346.1 | - | 6 - 7 | 734 | 61 | |
AK398108.1 | - | 6 - 7 | 752 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |