Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244343.1 | NP_001231272.1 | 3 - 4 | 198 | 78 | |
NM_001244344.1 | NP_001231273.1 | 2 - 3 | 336 | 78 | ||
XM_021100356.1 | XP_020956015.1 | 324 | 78 | |||
GenBank mRNA | AK351720.1 | - | 3 - 4 | 197 | 78 | |
BP161362.1 | - | 3 - 4 | 198 | 78 | ||
FS701378.1 | - | 2 - 3 | 336 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |