Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244347.1 | NP_001231276.1 | 1 - 2 | 294 | 58 | |
XM_021068214.1 | XP_020923873.1 | 661 | 58 | |||
XM_021068217.1 | XP_020923876.1 | 661 | 58 | |||
GenBank mRNA | AK236200.1 | - | 1 - 2 | 296 | 58 | |
AK347431.1 | - | 1 - 2 | 296 | 58 | ||
AK391059.1 | - | 1 - 2 | 369 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |