Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244347.1 | NP_001231276.1 | 4 - 5 | 1080 | 89 | |
XM_021068214.1 | XP_020923873.1 | 1447 | 89 | |||
XM_021068215.1 | XP_020923874.1 | 957 | 89 | |||
XM_021068216.1 | XP_020923875.1 | 1208 | 89 | |||
XM_021068217.1 | XP_020923876.1 | 1447 | 89 | |||
GenBank mRNA | AK236200.1 | - | 4 - 5 | 1082 | 89 | |
AK236359.1 | - | 4 - 5 | 1080 | 89 | ||
AK240388.1 | - | 4 - 5 | 918 | 89 | ||
AK347431.1 | - | 4 - 5 | 1082 | 89 | ||
AK391059.1 | - | 4 - 5 | 1155 | 89 | ||
AK398702.1 | - | 4 - 5 | 919 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |