Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF1AY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244349.1 | NP_001231278.1 | 2 - 3 | 214 | 150 | |
XM_013991067.2 | XP_013846521.2 | 292 | 150 | |||
GenBank mRNA | AY609402.1 | - | 2 - 3 | 214 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |