Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244361.1 | NP_001231290.1 | 3 - 4 | 534 | 61 | |
GenBank mRNA | AB295597.2 | - | 2 - 3 | 189 | 61 | |
AK397291.1 | - | 3 - 4 | 547 | 61 | ||
DB802979.1 | - | 3 - 4 | 534 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |