Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244362.1 | NP_001231291.1 | 9 - 10 | 797 | 112 | |
XM_021063996.1 | XP_020919655.1 | 987 | 112 | |||
XM_021063997.1 | XP_020919656.1 | 915 | 112 | |||
GenBank mRNA | AK238334.1 | - | 9 - 10 | 797 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |