Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244382.2 | NP_001231311.1 | 3 - 4 | 491 | 92 | |
NM_001244383.2 | NP_001231312.2 | 3 - 4 | 420 | 92 | ||
XM_005671150.3 | XP_005671207.1 | 366 | 92 | |||
XM_005671151.3 | XP_005671208.1 | 441 | 92 | |||
XM_005671152.3 | XP_005671209.1 | 543 | 92 | |||
GenBank mRNA | AK239027.1 | - | 3 - 4 | 306 | 92 | |
AK239590.1 | - | 3 - 4 | 393 | 92 | ||
AK239697.1 | - | 3 - 4 | 352 | 92 | ||
AK389707.1 | - | 3 - 4 | 394 | 92 | ||
AK392804.1 | - | 3 - 4 | 442 | 92 | ||
AK395192.1 | - | 3 - 4 | 437 | 92 | ||
AY610005.1 | - | 3 - 4 | 370 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |