Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244382.2 | NP_001231311.1 | 7 - 8 | 1036 | 98 | |
NM_001244383.2 | NP_001231312.2 | 7 - 8 | 965 | 98 | ||
XM_005671150.3 | XP_005671207.1 | 911 | 98 | |||
XM_005671151.3 | XP_005671208.1 | 986 | 98 | |||
XM_005671152.3 | XP_005671209.1 | 1088 | 98 | |||
GenBank mRNA | AK239027.1 | - | 7 - 8 | 851 | 98 | |
AK239590.1 | - | 7 - 8 | 938 | 98 | ||
AK239697.1 | - | 7 - 8 | 897 | 98 | ||
AK389707.1 | - | 7 - 8 | 939 | 98 | ||
AK392804.1 | - | 7 - 8 | 987 | 98 | ||
AK395192.1 | - | 7 - 8 | 982 | 98 | ||
AK398770.1 | - | 7 - 8 | 1018 | 98 | ||
AY610005.1 | - | 7 - 8 | 915 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |