Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHITM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244382.2 | NP_001231311.1 | 8 - 9 | 1208 | 116 | |
NM_001244383.2 | NP_001231312.2 | 8 - 9 | 1137 | 116 | ||
XM_005671150.3 | XP_005671207.1 | 1083 | 116 | |||
XM_005671151.3 | XP_005671208.1 | 1158 | 116 | |||
XM_005671152.3 | XP_005671209.1 | 1260 | 116 | |||
GenBank mRNA | AK239027.1 | - | 8 - 9 | 1023 | 116 | |
AK239697.1 | - | 8 - 9 | 1069 | 116 | ||
AK389707.1 | - | 8 - 9 | 1111 | 116 | ||
AK392804.1 | - | 8 - 9 | 1159 | 116 | ||
AK395192.1 | - | 8 - 9 | 1154 | 116 | ||
AK398770.1 | - | 8 - 9 | 1190 | 116 | ||
AY610005.1 | - | 8 - 9 | 1087 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |