Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244386.1 | NP_001231315.1 | 3 - 4 | 416 | 99 | |
XM_021100359.1 | XP_020956018.1 | 1015 | 99 | |||
GenBank mRNA | AK344118.1 | - | 3 - 4 | 411 | 99 | |
AY609657.1 | - | 3 - 4 | 416 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |