Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244398.1 | NP_001231327.1 | 5 - 6 | 366 | 88 | |
GenBank mRNA | AK236324.1 | - | 5 - 6 | 366 | 88 | |
AK393624.1 | - | 5 - 6 | 399 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |