Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244414.2 | NP_001231343.2 | 6 - 7 | 647 | 101 | |
XM_021088484.1 | XP_020944143.1 | 612 | 101 | |||
XM_021088485.1 | XP_020944144.1 | 1400 | 101 | |||
XM_021088486.1 | XP_020944145.1 | 761 | 101 | |||
XM_021088487.1 | XP_020944146.1 | 723 | 101 | |||
XM_021088488.1 | XP_020944147.1 | 732 | 101 | |||
XM_021088490.1 | XP_020944149.1 | 729 | 101 | |||
GenBank mRNA | AK230648.1 | - | 6 - 7 | 615 | 101 | |
AK230826.1 | - | 6 - 7 | 632 | 101 | ||
AK230913.1 | - | 6 - 7 | 632 | 101 | ||
AK230930.1 | - | 6 - 7 | 630 | 101 | ||
AK230937.1 | - | 6 - 7 | 634 | 101 | ||
AK230963.1 | - | 6 - 7 | 650 | 101 | ||
AK343772.1 | - | 6 - 7 | 647 | 101 | ||
AK347640.1 | - | 12 - 13 | 1400 | 101 | ||
AK389623.1 | - | 6 - 7 | 632 | 101 | ||
AK389775.1 | - | 6 - 7 | 615 | 101 | ||
AK389788.1 | - | 6 - 7 | 646 | 101 | ||
AK389799.1 | - | 6 - 7 | 647 | 101 | ||
AK389825.1 | - | 6 - 7 | 766 | 101 | ||
AK391408.1 | - | 6 - 7 | 650 | 101 | ||
AK399467.1 | - | 6 - 7 | 615 | 101 | ||
AK399483.1 | - | 6 - 7 | 635 | 101 | ||
AY609570.1 | - | 6 - 7 | 683 | 101 |
Gene Symbol: PLXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK389643.1 | - | 619 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |