Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244442.2 | NP_001231371.2 | 2 - 3 | 815 | 89 | |
GenBank mRNA | BX923393.2 | - | 1 - 2 | 131 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |