Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSFL1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244443.1 | NP_001231372.1 | 6 - 7 | 694 | 77 | |
XM_005672807.3 | XP_005672864.1 | 768 | 77 | |||
XM_005672808.3 | XP_005672865.1 | 619 | 77 | |||
GenBank mRNA | AK232184.1 | - | 6 - 7 | 675 | 77 | |
AK390042.1 | - | 7 - 8 | 768 | 77 | ||
AK392495.1 | - | 6 - 7 | 664 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |