Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244457.1 | NP_001231386.1 | 2 - 3 | 278 | 114 | |
XM_013982592.2 | XP_013838046.2 | 305 | 114 | |||
GenBank mRNA | AK232156.1 | - | 2 - 3 | 272 | 114 | |
AK344972.1 | - | 2 - 3 | 293 | 114 | ||
AK345556.1 | - | 2 - 3 | 279 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |