Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244501.2 | NP_001231430.1 | 9 - 10 | 1408 | 92 | |
GenBank mRNA | AK234901.1 | - | 9 - 10 | 1475 | 92 | |
AK235266.1 | - | 4 - 5 | 404 | 92 | ||
AK235921.1 | - | 9 - 10 | 1083 | 92 | ||
AK343698.1 | - | 6 - 7 | 656 | 92 | ||
AK392294.1 | - | 9 - 10 | 1408 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |