Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244522.1 | NP_001231451.1 | 1 - 2 | 389 | 68 | |
XM_005662047.3 | XP_005662104.2 | 474 | 68 | |||
GenBank mRNA | AK235025.1 | - | 1 - 2 | 389 | 68 | |
AK395742.1 | - | 1 - 2 | 393 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |