Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244522.1 | NP_001231451.1 | 4 - 5 | 674 | 85 | |
XM_005662047.3 | XP_005662104.2 | 759 | 85 | |||
GenBank mRNA | AK235025.1 | - | 4 - 5 | 674 | 85 | |
AK395742.1 | - | 4 - 5 | 678 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |